Rotors and Accessories for Eppendorf®, Rotor Adapter, Multipurpose Centrifuges 5804, 5804R, 5810 and 5810R
Supplier: Eppendorf
022638614 022638657 022638661 022638653 022637002 022637665 022637401 022637509 022637525 022637541 022637584 022637606 022637622 022637649 022637681 022637703 022637720 022638033 022637428 022637461 022637614 5820712007 5820715006 5820716002 022638505 022638564 022638866 022637436 022637100 022637207 022637568 022637452 022637673 022638840 022637436 022639905 5820742003 5820741007 5820747005 022638937 5820751002 5820708000 5820756004 022639907 5804747003 022639909 022639911 022639913 5920700006 5920701002 022638935 5920717006 022638084 022637479 022638556 022638614 022638629 022662970 5820761008 5825719000
CA022638614PK 3481.92 CAD
CA022638614 CA022638657 CA47743-070 CA47743-074 CA53513-816 CA53513-834 CA53513-840 CA53513-844 CA53513-846 CA53513-848 CA53513-852 CA53513-854 CA53513-856 CA53513-858 CA53513-860 CA53513-862 CA53513-864 CA53513-868 CA53513-904 CA53574-220 CA53574-224 CA5820711000 CA5820712007 CA5820713003 CA5820715006 CA5820716002 CA77888-208 CA77888-214 CA80078-568 CA89030-294 CA53513-818 CA53513-820 CA53513-850 CA53513-871 CA53574-222 CA80078-570 89030-294 CA11017-376 CA11017-384 CA11017-386 CA11017-396 CA11017-398 CA11017-400 CA11017-424 CA11017-426 CA11017-430 CA11017-432 CA11017-456 CA11017-460 CA11017-464 75874-870 75874-872 75874-902 75874-922 75874-924 75874-988 75875-110 75875-112 75875-120 75875-122 75875-124 75875-126 75875-136 75875-168 75875-172 75875-178
Rotors and Accessories for Eppendorf®, Rotor Adapter, Multipurpose Centrifuges 5804, 5804R, 5810 and 5810R
Centrifuges Rotors
Various adapters, rotors, and other accessories for centrifuges 5804 and/or 5804R and/or 5810 and/or 5810R. Eppendorf Rotors for Centrifuges 5804(R) and 5810(R) precision meets power. Customize your centrifugation with varied capacities and speeds. Accessorize with versatile buckets and adaptors for efficient sample processing. Ideal for high-performance applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight options available
Experience precision in high-performance centrifugation with Eppendorf Rotors designed for Centrifuge 5804/5804 R. These rotors offer diverse capacities and speeds, providing flexibility for various sample processing needs. Accessorize with versatile buckets and adaptors, enhancing the adaptability of your centrifuge setup. Ideal for high-performance applications, the Centrifuge 5804 series is further optimized with these precision-engineered rotors, ensuring accuracy and reproducibility in every spin. Explore the range to elevate your centrifugation experience.
Ordering information: Rotors, adapters, and accessories are only for use with centrifuge models specified. Unless otherwise specified, rotors may be used with all centrifuge models.