Admittance and Security Danger Signs, Construction and Maintenance, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
D470AC D470AD D470RC D470RD D496AB D496PB D496RB ESD132AB ESD132PB ESD132RB ESD266PB ESD266PC ESD266RB ESD266RC SPD450AB SPD450PB SPD450RB SPD59AB SPD59PB SPD59RB
94010-240EA 146.4 CAD
94010-240 94010-242 94010-244 94010-246 94010-410 94010-412 94010-414 94013-304 94013-306 94013-308 94013-392 94013-394 94013-396 94013-398 94021-624 94021-626 94021-628 94021-654 94021-656 94021-658
Admittance and Security Danger Signs, Construction and Maintenance, National Marker
Signs
Alert visitors and guest of security policies and direct them towards appropriate entrances
  • Available in Alum, PS vinyl, Rigid plastic (thick polystyrene), Glow polyester and Glow rigid plastic materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes
  • .010 glow polyester film and .050 glow rigid plastic materials are non-toxic, non-radioactive and glows for more than 6 hours
Each sign header reads "Danger".