Vileda Professional® Controlled Environment Trolleys
Supplier: Vileda Professional - FHP
141926 143774 149105
10146-008CS 8710.62 CAD
10146-008 10153-168 10769-306
Vileda Professional® Controlled Environment Trolleys
Mops
The CE trolley range has been specifically designed and developed for use in controlled environments where maximum contamination control is needed.
  • Two stainless steel mop handle holders
  • Support bar for Duo Press Wringer or Dip-N-Go Sieve, Support Plate and commonly used industry wringers
  • Frame holders allow for storage of various sized mops
  • Autoclavable, extremely quiet cleanroom casters
  • Electro-polished for a contamination free surface
  • Color coded system within boxes, buckets and handles
The trolleys are electro-polished stainless steel with air tight connections and smooth welding's
The CE Pre-Prepared Trolley features a top shelf working station, a storage area for Pre-Prepared mop box or 25L Longish bucket, and a 70L bin bag holder to dispose of used mops.
The CE Bucket Trolley features a rear area working station. A third bucket extension is available, turning double bucket system into triple bucket system.
Optional CE Third Bucket Extension attaches easily and securely to CE Bucket Trolley. Turns double bucket system into triple bucket system.
This ensures excellent biological control while still being easy to clean and full autoclavable. Trolleys offer ample space for all utensils needed for effective cleaning and are fully customizable to meet your exact needs.