PIPETBOY acu 2 Pipette Aid, INTEGRA Biosciences
Supplier: INTEGRA Biosciences
155019 155000 155015 155016 155017 155018
14222-180EA 1814.33 CAD
14222-180 37001-856 37001-858 37001-860 37001-862 37001-864
PIPETBOY acu 2 Pipette Aid, INTEGRA Biosciences
Pipettes Pipette Controllers
This second generation model, the PIPETBOY acu 2 Pipette Aid, is equipped with the latest technology to provide increased pipetting speed and a significant weight reduction in the instrument.
These two features help users to cut work time while offering an improved ergonomic pipetting experience. Besides these updates, the main benefits of the PIPETBOY acu Pipette Aid were preserved, including the unmatched pipetting precision, the colorful look, and the weight balance. Lighter and faster, the world’s most popular pipettor just got better.
Ordering information: PIPETBOY acu 2 Pipette Aids are supplied complete with Li-Ion battery, mains adapter, spare sterile 0.45 µm filter, and wall mount. A free Swiss pocket tool is included with each new PIPETBOY acu 2.