ASTM Liquid-in-Glass Mercury Thermometers, Total Immersion, Nos. 68 to 136, Thermco Products
Supplier: Thermco
ACC068C ACC068F ACC069C ACC069F ACC070C ACC070F ACC077F ACC078F ACC079F ACC080F ACC081F ACC097C ACC097F ACC098C ACC098F ACC108F ACC109F ACC110F ACC111C ACC113C ACC113F ACC114C ACC116C ACC117C ACC118C ACC118F ACC120C ACC121C ACC123C ACC124C ACC125C ACC126C ACC126F ACC127C ACC128C ACC128F ACC129C ACC129F ACC130C ACC130F ACC132C ACC136F
10747-226EA 508.95 CAD
10747-226 10747-228 10747-230 10747-232 10747-234 10747-236 10747-242 10747-244 10747-258 10747-260 10747-262 10747-264 10747-266 10747-282 10747-284 10747-286 10747-288 10747-310 10747-312 10747-314 10747-316 10747-318 10747-320 10747-322 10747-324 10747-326 10747-332 10747-334 10747-336 10747-338 10747-344 10747-346 10747-350 10747-352 10747-354 10747-356 10747-358 10747-360 10747-362 10747-364 10747-366 10747-368 10747-370 10747-372 10747-374 10747-384 10747-386
ASTM Liquid-in-Glass Mercury Thermometers, Total Immersion, Nos. 68 to 136, Thermco Products
Thermometers Analog Thermometers
These high precision liquid-in-glass mercury thermometers are hand blown, which bear graduations, numbers and letters etched into the glass and filled with and inert gas above the mercury column
ASTM thermometers are used for a variety of specified tests which quantify and qualify repeatable test results in the fields of oil and petroleum testing.
These thermometers were designed for specific testing and can be calibrated to your specific requirements.
Ordering information: Accessories not included. Certificate of calibration not included. ASTM thermometers with a NIST Traceable Certificate of Calibration from Thermco Products ISO/IEC accredited laboratory are also available; contact your Avantor representative for more information.
Caution: WARNING—Mercury has been designated by EPA and many state agencies as a hazardous material that can cause central nervous system, kidney and liver damage. Mercury, or its vapor, may be hazardous to health and corrosive to materials. Caution should be taken when handling mercury and mercury containing products. See the applicable product Safety Data Sheet (SDS) for details.