Hypersil GOLD™ HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
25002-052130-V 25002-102130-V 25002-104630 25002-152130-V 25003-034630 25002-052130
10041-002EA 1809.07 CAD
10041-002 10041-004 10041-006 10041-008 10041-012 10041-014 10041-022 10041-024 10041-026 10041-028 10041-034 10041-042 10041-058 10041-060 10041-062 10041-064 10041-084 10041-086 10045-220 10045-224 10045-232 10045-240 10045-242 10045-244 10045-258 10045-260 10045-262 10045-270 10045-272 10045-292 10045-306 10045-308 10045-310 10045-314 10045-316 10045-318 10045-322 10045-324 10045-326 10045-350 10045-366 10045-368 10045-384 10045-386 10045-390 10045-406 10045-408 10045-410 10045-412 10045-424 10045-428 10045-430 10045-432 10045-434 10045-444 10045-448 10045-452 10045-456 76449-768
Hypersil GOLD™ HPLC Columns, Thermo Scientific
Chromatography Columns
Achieve exceptional peak shape and resolution for your HPLC and LC/MS applications with Thermo Scientific Hypersil GOLD HPLC columns.
  • Endcapped, ultrapure, silica-based columns
  • Excellent resolution, efficiency, and sensitivity
  • Accuracy and quality of your analytical data
These endcapped, ultrapure, silica-based columns deliver significant reduction in peak tailing using generic gradients with C18 selectivity. With their excellent resolution, efficiency, and sensitivity, Hypersil GOLD columns give you confidence in the accuracy and quality of your analytical data. 1.9 µm particle size columns can improve the speed and efficiency of your applications.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD Holder, 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD Holder and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD Holder.