Paraffin Block Trimmer - Wax Trimmer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62850-50 62850-51
102094-478EA 122.45 CAD
102094-478 102094-480
Paraffin Block Trimmer - Wax Trimmer, Electron Microscopy Sciences
Paraffin Wax Trimmers and Dispensers
This Wax Trimmer is an appliance for removing excess paraffin from embedding tissue cassettes
  • Multiple blocks may be trimmed simultaneously
  • Grooved surface for proper wax drainage
  • Temperature settings of 80 and 90 °F
  • UL listed power cord
Its compact, tabletop design fits easily in any histology lab. Melted paraffin is shaved away. Collection tray stays in place by magnetic contact with the unit.
Delivery information: Includes 5 pieces disposable drip tray liner.