Uranyl zinc acetate

Supplier: ELECTRON MICROSCOPY SCIENCE CA
Danger

Synonyms: zinc bis(acetato-O)dioxouranate

22610 22610
CA10148-008EA 4763.16 CAD
101410-844 CA10148-008
Uranyl zinc acetate
Uranyl zinc acetate

Uranyl zinc acetate can be used as a laboratory reagent in the determination of sodium concentrations of solutions.

Formula: ZnUO₄(CH₃COO)₂
MW: 453.525 Da
Storage Temperature: Ambient
MDL Number: MFCD00036361
CAS Number: 10138-94-0
UN: 2910
ADR: 7,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR