Formalin 10%, TissuePrep, buffered
Supplier: Electron Microscopy Sciences
|
Danger
|
Synonyms:
Methylaldehyde
, Formol
, Paraformaldehyde aqueous solution
, Formaldehyde
Ready-to-use, prefilled with 10% formalin.
Prefilled with 10% formalin Cat# 15740. Ready-to-use in pre-filled cups. Labeled in accordance to OSHA requirements, which gives you immediate information regarding handling and first aid procedures. Pre-measured in wide-mouth, leak-proof, opaque polypropylene containers which limits your exposure to formaldehyde and saves you time.
Formula:
CH₂O MW: 30.03 g/mol |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...