Tissue Marking Dye Set, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

63020-BK 63020-BL 63020-RD 63020-GR 63020-YL 63020-04 63020-01 63020-OR 63020-PUR
100498-290EA 317.31 CAD
100498-290 100498-292 100498-296 100498-294 100498-298 100500-414 100498-288 102094-482 102094-484
Tissue Marking Dye Set, Electron Microscopy Sciences
Stains Histology Stains

Fast drying, easy-to-use dye that is ideal for marking frozen or fixed tissues.


  • Use with frozen or fixed tissues
  • Permanently marks margins of excised surgical specimens
  • Evenly coats the tissue without penetrating
  • Fast drying, easy-to-use


For use with frozen or fixed tissues.
Permanently mark the margins of excised surgical specimens for exact orientation.
Specially formulated binding pigment that evenly coats the tissue without penetrating the tissue surface.
The marking dyes do not inter


A specialized dye for marking tissue biopsies and pathology specimens.


Ordering information: Dyes are available individually (8 oz/bottle) or as a set of a 2 oz per bottle with the following colors: blue, black, red, yellow and green. The set also comes with 50 applicator sticks and a wooden storage case.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR