Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
11774-05
11774-10
11773-10
11772-05
11773-05
11772-10
100496-434EA
87.91
CAD
100496-434
100503-018
100503-016
100496-430
100496-432
100503-014
Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences
Buffers
Biological Buffers
All of our Dullbecco’s phosphate buffer saline solutions contain: potassium chloride, potassium phosphate monobasic, sodium chloride, sodium phosphate dibasic, and D-glucose.
- Some formulations may contain additional components
- Store at room temperature
Use the table below as a quick reference for identifying the one that best suits your application. All of the solutions are prepared 1X strength.