Paraformaldehyde, granular, EM Grade, purified

Supplier: Electron Microscopy Sciences
Danger

19210 19208
100504-162EA 89.9 CAD
100504-162 100504-160
Paraformaldehyde, granular, EM Grade, purified
Paraformaldehyde

Paraformaldehyde is the condensation product of methylene glycol, a substance not so far isolated in its pure form; it is a white solid with a formaldehyde content between 78 and 98% and is supplied either as a Prill or Granular.


All forms contain more than 95.5% paraformaldehyde content. The Prill and Granular forms are small, free-flowing, dust-free beads, the most desired by EM users.
A fast-penetrating EM fixative used in combination with Glutaraldehyde, Acrolein, and Osmium Tetroxide.
Karnovsky, M.J., J. Cell Biol., 27, 137A (1965). Hayat, M.A. Principles & Tech., of EM (Univ. Park Press), 68 (1981).

Formula: (CH₂O)n
Melting Pt: 120 °C (248 °F)
MDL Number: MFCD00133991
CAS Number: 30525-89-4
UN: 2213
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f240682c03df85e943acbb1a983fe71750db44cd&vid=96c37abe2cf4c6160dda3947fb05d9d23c32fa74&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR