Formalin 10% (w/v), acetate buffered (neutral)

Supplier: Electron Microscopy Sciences
Warning

Synonyms: MethylaldehydeFormolParaformaldehyde aqueous solutionFormaldehyde

15738-01 15738-04
100496-498EA 84.2 CAD
100496-498 100496-500
Formalin 10% (w/v), acetate buffered (neutral)
Formalin

Buffered with sodium acetate in accordance with the A.F.I.P. manual of histology.


  • Filtered
  • Ready-to-use

Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: 100 °C (212 °F)
Density: 1 g/cm³ (8.345 lbs/gal.)
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 3334
ADR: 9,

Order Now

Specification Test Results

pH at 20°C 6.0 - 7.1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR