Formaldehyde/Zinc Fixative, Electron Microscopy Science

Supplier: Electron Microscopy Sciences

15675-04 15675-01
100496-488EA 209.92 CAD
100496-488 100496-486
Formaldehyde/Zinc Fixative, Electron Microscopy Science
Stains Microscopy and staining reagents

Formaldehyde/zinc fixative is ready-to-use solution. Can be used with automated tissue processors.


  • Preserves fine nuclear details
  • Good for immunostaining
  • Contains no mercury


Works well with standard and special stains because zinc is a stain enhancing mordant.

Order Now

SPECIFICATIONS

Environmentally Preferable
Storage temperature RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR