Formaldehyde 4% in aqueous solution, EM Grade

Supplier: Electron Microscopy Sciences
Danger

157-4 157-4-100
102091-904EA 95.46 CAD
102091-904 102091-906
Formaldehyde 4% in aqueous solution, EM Grade
Formaldehyde

Paraformaldehyde - methanol free solution.


A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives.


Since the DOT (Department of Transportation) has changed the definition of what a non dangerous solution of formaldehyde is to under 10%, we are now offering an 8% and a 4% solution. This will allow us to ship overnight as well as overseas without any hazardous charges saving our customers a good deal of money.


Packaging: Easy to break, prescored, 10 ml ampoules sealed under inert gas, or 100 ml bottles.

Formula: CH₂O
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f240682c03df85e943acbb1a983fe71750db44cd&vid=96c37abe2cf4c6160dda3947fb05d9d23c32fa74&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR