Fixatives, Prepared, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
15953 15956 15980 15949 15950 15970 15960 15960-01
100503-950EA 114.85 CAD
100503-950 100503-954 100503-960 100504-860 100504-862 100503-958 100503-956 MSPP-15960-01
Fixatives, Prepared, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Premixed, prebuffered fixatives, that are pH adjusted for immediate laboratory use. They are freshly prepared upon receipt of order to ensure maximum shelf life. The fixatives are filled in 20 mL glass vials (10 mL/vial). This allows for the immediate fixation of your specimens without costly and time consuming transfer associated with ampoule contained fixatives.
Certifications: Certificates of analysis are available.