Decalcifier, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
64147-10 64147-06 64147-01
100498-642EA 518.67 CAD
100498-642 100500-438 100498-640
Decalcifier, Electron Microscopy Science
Stains Microscopy and staining reagents
Protects cellular structure. Can be used with human and animal calcified tissue, bone, cartilage and other hard specimens.
  • Ready-to-use
  • Minimal interference with subsequent staining
Ready to use decalcifier protects cellular structure and causes minimal interference with subsequent staining. For use with human and animal calcified tissue, bone, cartilage and other hard specimens.