Formalin 10% in aqueous solution low odor, buffered

Supplier: Electron Microscopy Sciences
Warning

Synonyms: MethylaldehydeFormolParaformaldehyde aqueous solutionFormaldehyde

15741-04 15741-01
100500-246CS 719.82 CAD
100500-246 100496-506
Formalin 10% in aqueous solution low odor, buffered
Formalin

10% formalin in Phosphate buffer according to the A.F.I.P. Manual of Histology.


  • Low odor


This solution allows for long-term storage of tissue specimens.

Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: 100 °C (212 °F)
Density: 0.83 g/cm³ (6.926 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 3334
ADR: 9,
https://ca.vwr.com/stibo/web/std.lang.all/48/24/28984824.jpg

Order Now

Specification Test Results

pH at 25 °C 6.9 - 7.1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR