Potts-Smith Scissors, OR Grade, Sklar
Supplier: Sklar
98-1075 98-1076 52-2860 52-2845 52-2825
10184-952EA 1344.03 CAD
10184-952 10184-954 10194-308 10194-314 10199-416
Potts-Smith Scissors, OR Grade, Sklar
Scissors
Sklar's® Potts-Smith Scissors are a finger ring instrument commonly used for trimming and opening vessels in cardiovascular and thoracic procedures.
Blades are available in a 20 mm intermediate weight or 13 mm delicate weight style
Censitrac Ready products are industry standard and constructed of stainless steel.
Delicate Potts-Smith scissors are heavier than Diethrich scissors, and can therefore also be used for cutting through plaque.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for theMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable