Aufranc Cobra Retractor, Sklar®
Supplier: Sklar
40-8284 40-8280 40-8282
10194-012EA 525.96 CAD
10194-012 10194-416 10196-560
Aufranc Cobra Retractor, Sklar®
Retractors
Aufranc Retractor, which may also be known as a Cobra retractor, is an orthopedic instrument used in procedures involving large bones and joints
The blade of the retractor flares out and then narrows again at the tip, representing the shape of a cobra.