Volkman Retractor, OR Grade, Sklar®
Supplier: Sklar
60-5604 60-5504 60-4243 60-4244 60-4253 60-4254 60-4256 60-5502 60-5503 60-5506 60-5602 60-5603 60-5606 60-5812 60-5813 60-5814 60-5816 60-5820 60-5822 60-5823 60-5824 60-5826
10194-244EA 291.66 CAD
10194-244 10194-246 10197-756 10197-758 10197-760 10197-762 10197-764 10197-778 10197-780 10197-782 10197-784 10197-786 10197-788 10197-790 10197-792 10197-794 10197-796 10197-798 10197-800 10197-802 10197-804 10197-806
Volkman Retractor, OR Grade, Sklar®
Retractors
Sklar's® Volkman Retractor is a handheld retractor commonly used in small bone and joint procedures. The Volkman retractor is available with 1 to 6 prong tips and the prongs can be sharp or blunt, depending on the surgical site and tissue type being held.
This retractor may also be referred to as the Volkman Rake Retractor when there are multiple prongs
Censitrac Ready products are industry standard and constructed of stainless steel.
It features both USA style handle and standard style handle which are fenestrated circle and tear drop.
Ordering information: Censitrac instrumentsMore Product Information