Farr Retractor, OR Grade, Sklar®
Supplier: Sklar
22-8842 22-8839
10195-232EA 87.41 CAD
10195-232 10195-234 10195-236 76192-920 76192-922 76192-924
Farr Retractor, OR Grade, Sklar®
Retractors
The Farr Retractor is a self-retaining spring retractor, used to hold back tissue during abdominal and hernia procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard