Richardson-Eastman Retractor, OR Grade, Sklar®
Supplier: Sklar
60-1697 60-1699
10197-276PK 138.05 CAD
10197-276 10197-278 10197-280
Richardson-Eastman Retractor, OR Grade, Sklar®
Retractors
The Richardson-Eastman Double-Ended Retractor is a handheld, right angle retractor used for holding back layers of tissue close to the surface.
  • Available in different sizes with crescent shaped lip
  • Smaller blade for an initial retraction
  • Larger blade for full retraction
  • The Censitrac Ready products are industry standard
The large retractor has a long blade measures of 33/4" x 13/4" and a short blade that measures 13/4" x 11/2". The small retractor has a long blade measures 11/4" x 11/8" and a short blade measures 1" x 7/8".
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.