Merit™ Weitlaner Retractor, Physician Grade, Sklar
Supplier: Sklar
97-370 97-373 97-376 97-371 97-372
10184-426EA 210.18 CAD
10184-426 10184-428 10192-432 10193-972 75818-084
Merit™ Weitlaner Retractor, Physician Grade, Sklar
Retractors
Merit's™ Weitlaner Retractor is a self-retaining, finger ring retractor with a cam ratchet lock used for holding back tissue and exposing a surgical site
It has curved shanks that lead to blades that have between two and six, sharp or blunt, outward curving prongs. The prongs interdigitate when the instrument is closed. The Weitlaner retractor is a popular instrument, most commonly used in basic plastic surgery, large bone and joint procedures, and mastoid surgeries. It is also useful with small, deep incisions and soft tissue dissection at a superficial level. Merit™ instruments are Sklar’s office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit™ instruments deliver an economical solution for physicians and practices that require quality, durable instruments.