Desmarres Retractor, Smooth, OR Grade, Sklar®
Supplier: Sklar
65-1841 65-1842 65-1843 65-1844
10194-654EA 224.87 CAD
10194-654 10194-656 10194-658 10194-660
Desmarres Retractor, Smooth, OR Grade, Sklar®
Retractors
Desmarres Lid Retractor is a handheld ophthalmologic retractor used for retaining primarily the upper eyelid and eyelashes
It may be used to pull down the lower lid as well. The curved, saddle style blade contours to the round shape of the eye as well as the curvature of the eyelid created when it is retracted. It is available in multiple sizes to fit the patient's eye.