Anti-APBA1 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18702
CAPIPA5-18702EA 2904.29 CAD
CAPIPA5-18702
Anti-APBA1 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. The protein encoded by this gene is a member of the X11 protein family. It is a neuronal adapter protein that interacts with the Alzheimer's disease amyloid precursor protein . It stabilizes APP and inhibits production of proteolytic APP fragments including the A beta peptide that is deposited in the brains of Alzheimer's disease patients. This gene product is believed to be involved in signal transduction processes. It is also regarded as a putative vesicular trafficking protein in the brain that can form a complex with the potential to couple synaptic vesicle exocytosis to neuronal cell adhesion.

Recommended Dilutions: Western Blot: 0.1-0.3 ug/ml

Type: Primary
Antigen: APBA1 (amyloid beta (A4) precursor protein-binding, family A, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR