Anti-CGI58 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18666
CAPIPA5-18666EA 3704.55 CAD
CAPIPA5-18666
Anti-CGI58 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine and rat based on sequence homology. The protein encoded by this gene belongs to a large family of proteins defined by an alpha/beta hydrolase fold, and contains three sequence motifs that correspond to a catalytic triad found in the esterase/lipase/thioesterase subfamily. It differs from other members of this subfamily in that its putative catalytic triad contains an asparagine instead of the serine residue. Mutations in this gene have been associated with Chanarin-Dorfman syndrome, a triglyceride storage disease with impaired long-chain fatty acid oxidation.

Western Blot: 0.2-0.6 ug/ml

Type: Primary
Antigen: CGI58
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Human, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR