Anti-RNF213/C17orf27 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18650
CAPIPA5-18650EA 3417.45 CAD
CAPIPA5-18650
Anti-RNF213/C17orf27 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunohistochemistry: 2.5µg/ml

Type: Primary
Antigen: RNF213/C17orf27
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR