Anti-EPB41L3 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18453
CAPIPA5-18453EA 3269.49 CAD
CAPIPA5-18453
Anti-EPB41L3 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine, mouse and rat based on sequence homology. Critical growth regulator in the pathogenesis of meningiomas.

Recommended Dilutions: Western Blot: 1-3 ug/ml

Type: Primary
Antigen: EPB41L3 (erythrocyte membrane protein band 4.1-like 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR