Anti-SORD Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18333
CAPIPA5-18333EA 3269.49 CAD
CAPIPA5-18333
Anti-SORD Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine, human and rat based on sequence homology. Sorbitol dehydrogenase catalyzes the interconversion of polyols and their corresponding ketoses, and together with aldose reductase is the reduction of glucose to sorbitol by ALDR1 with NADPH as the cofactor. SORD then oxidizes the sorbitol to fructose using NADcofactor.

Recommended Dilutions: Western Blot: 0.1-0.3 ug/ml

Type: Primary
Antigen: SORD (sorbitol dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR