Anti-FNBP1 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18302
CAPIPA5-18302EA 3269.49 CAD
CAPIPA5-18302
Anti-FNBP1 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. The protein encoded by this gene is a member of the formin-binding-protein family. The protein contains an N-terminal Fer/Cdc42-interacting protein 4 homology domain followed by a coiled-coil domain, a proline-rich motif, a second coiled-coil domain, a Rho family protein-binding domain , and a C-terminal SH3 domain. This protein binds sorting nexin 2 , tankyrase , and dynamin; an interaction between this protein and formin has not been demonstrated yet in human.

Recommended Dilutions: Western Blot: 0.25-2 ug/ml

Type: Primary
Antigen: FNBP1 (formin binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR