Anti-ACTR1B Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18274
CAPIPA5-18274EA 3269.49 CAD
CAPIPA5-18274
Anti-ACTR1B Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This gene encodes a 42.3 kD subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein and is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit, like ACTR1A, is an actin-related protein. These two proteins, which are of equal length and share 90% amino acid identity, are present in a constant ratio of approximately 1:15 in the dynactin complex.

Immunohistochemistry: 3-5µg/ml, Western Blot: 0.5-2 ug/ml

Type: Primary
Antigen: ACTR1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR