Anti-APOM Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18980
CAPIPA5-18980EA 3269.49 CAD
CAPIPA5-18980
Anti-APOM Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. The protein encoded by this gene is an apolipoprotein and member of the lipocalin protein family. It is found associated with high density lipoproteins and to a lesser extent with low density lipoproteins and triglyceride-rich lipoproteins. The encoded protein is secreted through the plasma membrane but remains membrane-bound, where it is involved in lipid transport. Two transcript variants encoding two different isoforms have been found for this gene, but only one of them has been fully characterized.

Western Blot: 0.3-1 ug/ml

Type: Primary
Antigen: APOM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR