Anti-FZD2 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18884
CAPIPA5-18884EA 3199.34 CAD
CAPIPA5-18884
Anti-FZD2 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. This intronless gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the wingless type MMTV integration site family of signaling proteins. This gene encodes a protein that is coupled to the beta-catenin canonical signaling pathway. Competition between the wingless-type MMTV integration site family, member 3A and wingless-type MMTV integration site family, member 5A gene products for binding of this protein is thought to regulate the beta-catenin-dependent and -independent pathways.

Recommended Dilutions: Immunohistochemistry: 2-5µg/ml

Type: Primary
Antigen: FZD2 (frizzled class receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR