Anti-FZD7 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18883
CAPIPA5-18883EA 2904.29 CAD
CAPIPA5-18883
Anti-FZD7 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse based on sequence homology. Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD7 protein contains an N-terminal signal sequence, 10 cysteine residues typical of the cysteine-rich extracellular domain of Fz family members, 7 putative transmembrane domains, and an intracellular C-terminal tail with a PDZ domain-binding motif. FZD7 gene expression may downregulate APC function and enhance beta-catenin-mediated signals in poorly differentiated human esophageal carcinomas.

Recommended Dilutions: Western Blot: 0.3-1 ug/ml

Type: Primary
Antigen: FZD7 (frizzled class receptor 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR