Anti-ATP13A1 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18860
CAPIPA5-18860EA 3268.55 CAD
CAPIPA5-18860
Anti-ATP13A1 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine, mouse and rat based on sequence homology.

Western Blot: 0.3-1 ug/ml

Type: Primary
Antigen: ATP13A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR