Anti-ITGAM Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18727
CAPIPA5-18727EA 2904.29 CAD
CAPIPA5-18727
Anti-ITGAM Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This gene encodes the integrin alpha M chain. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This I-domain containing alpha integrin combines with the beta 2 chain to form a leukocyte-specific integrin referred to as macrophage receptor 1 receptor 3 . The alpha M beta 2 integrin is important in the adherence of neutrophils and monocytes to stimulated endothelium, and also in the phagocytosis of complement coated particles. Multiple transcript variants encoding different isoforms have been found for this gene.

Immunohistochemistry: 2-4 ug/ml, Western Blot: 1-3 ug/ml

Type: Primary
Antigen: ITGAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR