Anti-SALL4 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18717
CAPIPA5-18717EA 3269.49 CAD
CAPIPA5-18717
Anti-SALL4 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse based on sequence homology.

Recommended Dilutions: Western Blot: 0.1-0.3 ug/ml

Type: Primary
Antigen: SALL4 (spalt-like transcription factor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR