Anti-PDE4D3 Goat Polyclonal Antibody

Supplier: Avantor
CAPIPA5-18621EA 3175.89 CAD
CAPIPA5-18621
Anti-PDE4D3 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with human and porcine based on sequence homology. This gene encodes one of four mammalian counterparts to the fruit fly 'dunce' gene. The encoded protein has 3',5'-cyclic-AMP phosphodiesterase activity and degrades cAMP, which acts as a signal transduction molecule in multiple cell types. This gene uses different promoters to generate multiple alternatively spliced transcript variants that encode functional proteins.

Recommended Dilutions: Western Blot: 0.01-0.03 ug/ml

Type: Primary
Antigen: PDE4D3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR