Anti-Nur77/ TR3 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18573
CAPIPA5-18573EA 3269.49 CAD
CAPIPA5-18573
Anti-Nur77/ TR3 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This gene encodes a member of the steroid-thyroid hormone-retinoid receptor superfamily. Expression is induced by phytohemagglutinin in human lymphocytes and by serum stimulation of arrested fibroblasts. The encoded protein acts as a nuclear transcription factor. Translocation of the protein from the nucleus to mitochondria induces apoptosis. Multiple transcript variants encoding different isoforms have been found for this gene.

Recommended Dilutions: Western Blot: 1-3 ug/ml

Type: Primary
Antigen: Nur77/ TR3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR