Anti-PDE4D Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18459
CAPIPA5-18459EA 3417.45 CAD
CAPIPA5-18459
Anti-PDE4D Goat Polyclonal Antibody
Antibodies
This gene encodes one of four mammalian counterparts to the fruit fly 'dunce' gene. The encoded protein has 3',5'-cyclic-AMP phosphodiesterase activity and degrades cAMP, which acts as a signal transduction molecule in multiple cell types. This gene uses different promoters to generate multiple alternatively spliced transcript variants that encode functional proteins.

Recommended Dilutions: Western Blot: 0.1-0.3 ug/ml, ELISA: Assay-dependent

Type: Primary
Antigen: PDE4D (phosphodiesterase 4D, cAMP-specific)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR