Anti-VAV3 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18296
CAPIPA5-18296EA 2841.96 CAD
CAPIPA5-18296
Anti-VAV3 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine based on sequence homology. This gene is a member of the VAV gene family. The VAV proteins are guanine nucleotide exchange factors for Rho family GTPases that activate pathways leading to actin cytoskeletal rearrangements and transcriptional alterations. This gene product acts as a GEF preferentially for RhoG, RhoA, and to a lesser extent, RAC1, and it associates maximally with the nucleotide-free states of these GTPases. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.

Recommended Dilutions: Western Blot: 0.3-1 ug/ml

Type: Primary
Antigen: VAV3 (vav 3 guanine nucleotide exchange factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR