Anti-SMUG1 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18262
CAPIPA5-18262EA 3269.49 CAD
CAPIPA5-18262
Anti-SMUG1 Goat Polyclonal Antibody
Antibodies
SMUG1 is a glycosylase that removes uracil from single- and double-stranded DNA in nuclear chromatin, thus contributing to base excision repair.

Recommended Dilutions: Western Blot: 0.5-2 ug/ml

Type: Primary
Antigen: SMUG1 (single-strand-selective monofunctional uracil-DNA glycosylase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR