Anti-NDC80 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18089
CAPIPA5-18089EA 3269.49 CAD
CAPIPA5-18089
Anti-NDC80 Goat Polyclonal Antibody
Antibodies
HEC is one of several proteins involved in spindle checkpoint signaling. This surveillance mechanism assures correct segregation of chromosomes during cell division by detecting unaligned chromosomes and causing prometaphase arrest until the proper bipolar attachment of chromosomes is achieved.

  • Recommended Dilutions: Western Blot: 0.5-2 ug/ml

Type: Primary
Antigen: NDC80 (NDC80 kinetochore complex component)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR