Anti-RAB11A/YL8 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18081
CAPIPA5-18081EA 2904.29 CAD
CAPIPA5-18081
Anti-RAB11A/YL8 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, human and rat based on sequence homology. The protein encoded by this gene belongs to the small GTPase superfamily, Rab family. It is associated with both constitutive and regulated secretory pathways, and may be involved in protein transport.

Recommended Dilutions: Western Blot: 1-3 ug/ml

Type: Primary
Antigen: RAB11A/YL8
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR