Anti-BPOZ Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-17974
CAPIPA5-17974EA 3269.49 CAD
CAPIPA5-17974
Anti-BPOZ Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This gene encodes a protein with an ankyrin repeat region and two BTB/POZ domains, which are thought to be involved in protein-protein interactions. Expression of this gene is activated by the phosphatase and tensin homolog, a tumor suppressor. Alternate splicing results in three transcript variants.

Immunohistochemistry: 3-5µg/ml, Western Blot: 1-3 ug/ml

Type: Primary
Antigen: BPOZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR