EN Fire - Rated Safety Cabinets, Justrite®
Supplier: Justrite
22601 22603 22605 22607
10803-740EA 10569.81 CAD
10803-740 10803-742 10803-744 10803-746
EN Fire - Rated Safety Cabinets, Justrite®
Furniture Storage Cabinets
Safely store and segregate flammables in either 30- or 90-minute Fire-Rated Safety Cabinets.
  • Hybrid-close door action; manual close doors stop in any position, but self-close in the event of a fire to protect contents
  • Cabinets are supplied with 3 adjustable shelves
Certifications: FM, EN 14470-1, EN 14727, NFPA 30, OSHA, and NFPA 1 - 2012 Edition.