Merit™ Rochester-Carmalt Forceps, Physician Grade, Sklar
Supplier: Sklar
97-534 97-537
10184-854EA 43.38 CAD
10184-854 10184-856
Merit™ Rochester-Carmalt Forceps, Physician Grade, Sklar
Forceps
Merit™ Rochester-Carmalt Hemostatic Forceps are a standard type of hemostatic clamp used in several procedures.
These forceps have longitudinal serrations the entire length of the jaw. The tips are cross-serrated. The fully serrated working surface allows this instrument to have greater grip on heavy or tough tissue and stop blood flow of large vessels. The ratcheted handle allows locking on larger tissue for hemostasis.
Merit™ instruments are office-grade products used for routine and in-office procedures. These high quality instruments are crafted from Pakistan steel with a 10 year guarantee. Merit™ instruments deliver a solution for physicians and practices that require quality, durable instruments.