Merit™ Rankin-Kelly Hemostatic Forceps, Physician Grade, Sklar
Supplier: Sklar
97-465 97-469
10184-810EA 48.43 CAD
10184-810 10184-812
Merit™ Rankin-Kelly Hemostatic Forceps, Physician Grade, Sklar
Forceps
Merit™ Rankin-Kelly Forceps are used for clamping large blood vessels or manipulating heavy tissue
They are available curved or straight with a ratcheted finger ring handle. Rankin and Kelly forceps are almost identical. The jaws of both are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws, however, Rankin forceps are longer and lighter than Kelly forceps.
Merit™ instruments are mid-grade products used for routine and in-office procedures. These high quality instruments are crafted from Pakistan steel with a 10 year guarantee. Merit™ instruments deliver a solution for physicians and practices that require quality, durable instruments.
They may also be used for soft tissue dissection.