Castroviejo Fixation Forceps, OR Grade, Sklar
Supplier: Sklar
66-2635 66-2640
10197-484EA 666.18 CAD
10197-484 10197-486
Castroviejo Fixation Forceps, OR Grade, Sklar
Forceps
Castroviejo Fixation Forceps are ophthalmologic thumb forceps used to hold tissue in place before incisions.
They are a ratcheted instrument, which provides a secure grip on the tissue, so that it does not move.